Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142950.1 | NP_001136422.1 | 2 - 3 | 219 | 84 | |
NM_027350.3 | NP_081626.2 | 2 - 3 | 216 | 84 | ||
GenBank mRNA | AK077699.1 | - | 2 - 3 | 197 | 84 | |
AK083730.1 | - | 2 - 3 | 176 | 84 | ||
AK157244.1 | - | 2 - 3 | 178 | 84 | ||
AK172657.1 | - | 2 - 3 | 178 | 84 | ||
AK191424.1 | - | 1 - 2 | 37 | 84 | ||
BC005473.1 | - | 2 - 3 | 84 | |||
BC052849.1 | - | 2 - 3 | 177 | 30057822 | 84 | |
BY117114.1 | - | 2 - 3 | 220 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |