Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142950.1 | NP_001136422.1 | 6 - 7 | 552 | 65 | |
NM_027350.3 | NP_081626.2 | 6 - 7 | 549 | 65 | ||
GenBank mRNA | AK077699.1 | - | 6 - 7 | 530 | 65 | |
AK172657.1 | - | 6 - 7 | 511 | 65 | ||
AW545712.1 | - | 4 - 5 | 65 | |||
BC005473.1 | - | 6 - 7 | 65 | |||
BC052849.1 | - | 6 - 7 | 510 | 30057822 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |