Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008540.2 | NP_032566.2 | 6 - 7 | 1224 | 75 | |
GenBank mRNA | AB221585.1 | - | 5 - 6 | 784 | 75 | |
AK004804.1 | - | 5 - 6 | 588 | 75 | ||
AY493561.1 | - | 5 - 6 | 784 | 75 | ||
BC046584.1 | - | 6 - 7 | 1224 | 6313280 | 75 | |
GU072920.1 | - | 5 - 6 | 784 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |