Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289648.1 | NP_001276577.1 | 7 - 8 | 675 | 81 | |
NM_001289649.1 | NP_001276578.1 | 7 - 8 | 654 | 81 | ||
NM_001289650.1 | NP_001276579.1 | 7 - 8 | 689 | 81 | ||
NM_029019.4 | NP_083295.1 | 7 - 8 | 744 | 81 | ||
GenBank mRNA | AF480303.1 | - | 4 - 5 | 391 | 81 | |
AK014759.1 | - | 7 - 8 | 655 | 81 | ||
AK016984.1 | - | 7 - 8 | 655 | 81 | ||
BC061022.1 | - | 5 - 6 | 427 | 6774873 | 81 | |
BC087893.1 | - | 7 - 8 | 666 | 6774832 | 81 | |
JF440953.1 | - | 5 - 6 | 463 | 81 | ||
JF440954.1 | - | 5 - 6 | 463 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |