Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085373.1 | NP_001078842.1 | 14 - 15 | 2188 | 85 | |
NM_001085374.1 | NP_001078843.1 | 12 - 13 | 2156 | 85 | ||
GenBank mRNA | AK158266.1 | - | 14 - 15 | 2189 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |