Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085373.1 | NP_001078842.1 | 17 - 18 | 2724 | 54 | |
NM_001085374.1 | NP_001078843.1 | 15 - 16 | 2692 | 54 | ||
GenBank mRNA | AK054528.1 | - | 15 - 16 | 2695 | 54 | |
AK158266.1 | - | 17 - 18 | 2725 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |