Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019514.3 | NP_062387.2 | 8 - 9 | 1754 | 92 | |
NM_207109.2 | NP_996992.1 | 9 - 10 | 1910 | 92 | ||
GenBank mRNA | AF116573.1 | - | 6 - 7 | 922 | 92 | |
AK041160.1 | - | 10 - 11 | 1814 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |