Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037711.2 | NP_001032800.2 | 9 - 10 | 1902 | 66 | |
NM_001293727.1 | NP_001280656.1 | 9 - 10 | 1878 | 66 | ||
GenBank mRNA | AK173148.1 | - | 9 - 10 | 1879 | 66 | |
BC042459.1 | - | 9 - 10 | 1848 | 66 | ||
BC064474.1 | - | 9 - 10 | 1826 | 66 | ||
BC108378.1 | - | 1862 | 30736765 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |