Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ajap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099299.1 | NP_001092769.1 | 2 - 3 | 1507 | 85 | |
GenBank mRNA | BC147229.1 | - | 2 - 3 | 871 | 9056280 | 85 |
BC147230.1 | - | 2 - 3 | 871 | 9056273 | 85 | |
BC147654.1 | - | 2 - 3 | 871 | 9087443 | 85 | |
BC147658.1 | - | 2 - 3 | 871 | 9087447 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |