Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ajap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099299.1 | NP_001092769.1 | 4 - 5 | 1838 | 114 | |
GenBank mRNA | BC147229.1 | - | 4 - 5 | 1202 | 9056280 | 114 |
BC147230.1 | - | 4 - 5 | 1202 | 9056273 | 114 | |
BC147654.1 | - | 4 - 5 | 1202 | 9087443 | 114 | |
BC147658.1 | - | 4 - 5 | 1202 | 9087447 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |