Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160016.1 | NP_001153488.1 | 1 - 2 | 218 | 80 | |
NM_008142.4 | NP_032168.1 | 1 - 2 | 218 | 80 | ||
GenBank mRNA | AK163844.1 | - | 1 - 2 | 207 | 80 | |
BC013058.1 | - | 1 - 2 | 209 | 3964965 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |