Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021303.2 | NP_067278.2 | 7 - 8 | 820 | 69 | |
GenBank mRNA | AK076287.1 | - | 6 - 7 | 862 | 69 | |
AK078560.1 | - | 7 - 8 | 796 | 69 | ||
AK133834.1 | - | 7 - 8 | 820 | 69 | ||
AK157765.1 | - | 7 - 8 | 813 | 69 | ||
AK184541.1 | - | 3 - 4 | 265 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |