Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183300.2 | NP_899123.2 | 4 - 5 | 517 | 60 | |
GenBank mRNA | DQ303413.1 | - | 4 - 5 | 517 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |