Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172872.3 | NP_766460.2 | 6 - 7 | 2545 | 78 | |
GenBank mRNA | AK081525.1 | - | 6 - 7 | 2483 | 78 | |
BC060737.1 | - | 6 - 7 | 2559 | 4191502 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |