Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163767.1 | NP_001157239.1 | 1 - 2 | 208 | 63 | |
GenBank mRNA | AK006804.1 | - | 1 - 2 | 209 | 63 | |
BC048635.1 | - | 1 - 2 | 180 | 63 | ||
BC099549.1 | - | 1 - 2 | 214 | 30919464 | 63 | |
BC132398.1 | - | 1 - 2 | 158 | 40130675 | 63 | |
BC132400.1 | - | 1 - 2 | 158 | 40130677 | 63 | |
BC145187.1 | - | 1 - 2 | 158 | 9053738 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |