Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003917.4 | NP_001003917.2 | 10 - 11 | 1869 | 66 | |
NM_001288612.1 | NP_001275541.1 | 10 - 11 | 1872 | 66 | ||
NM_001288613.1 | NP_001275542.1 | 10 - 11 | 2117 | 66 | ||
NR_109938.1 | 10 - 11 | 1813 | 66 | |||
GenBank mRNA | AM085510.1 | - | 10 - 11 | 1782 | 66 | |
BC034139.1 | - | 3 - 4 | 558 | 66 | ||
BC056488.1 | - | 10 - 11 | 2030 | 66 | ||
BC079884.1 | - | 10 - 11 | 1758 | 6840050 | 66 | |
BC082307.1 | - | 8 - 9 | 1575 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |