Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162950.1 | NP_001156422.1 | 8 - 9 | 1068 | 88 | |
NM_016868.3 | NP_058564.2 | 8 - 9 | 1099 | 88 | ||
GenBank mRNA | AB289606.1 | - | 8 - 9 | 1035 | 88 | |
AF060194.1 | - | 8 - 9 | 1099 | 88 | ||
AK144472.1 | - | 3 - 4 | 442 | 88 | ||
BC120587.1 | - | 8 - 9 | 1052 | 40129510 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |