Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146200.1 | NP_001139672.1 | 4 - 5 | 2432 | 139 | |
NM_001146201.1 | NP_001139673.1 | 4 - 5 | 2488 | 139 | ||
NM_020272.2 | NP_064668.2 | 4 - 5 | 2609 | 139 | ||
GenBank mRNA | AF208345.1 | - | 3 - 4 | 2288 | 139 | |
AJ249280.1 | - | 4 - 5 | 2609 | 139 | ||
AK040543.1 | - | 4 - 5 | 2394 | 139 | ||
AK132553.1 | - | 4 - 5 | 2434 | 139 | ||
AK137470.1 | - | 4 - 5 | 2341 | 139 | ||
AK137620.1 | - | 4 - 5 | 2490 | 139 | ||
AK143238.1 | - | 1 - 2 | 113 | 139 | ||
BC051246.1 | - | 4 - 5 | 2349 | 6514979 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |