Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm41764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078684.1 | - | 245 | 114 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | low density lipoprotein receptor class A domain containing 4 | Ldlrad4 |
8230401C20Rik, A430083H02, A430108L08Rik, C18orf1, D18Ertd653e, D330030L18Rik |
Anti-sense | Chr. 18 - 67932245 - 68260550 on NCBI Build GRCm38 | |
Downstream |
family with sequence similarity 210, member A |
Fam210a |
4933403F05Rik, AA987140, AV134576, AW491926 |
155.9kb | Sense | Chr. 18 - 68260184 - 68300333 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |