Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008233.3 | NP_001008234.2 | 7 - 8 | 776 | 79 | |
GenBank mRNA | AK048084.1 | - | 5 - 6 | 2755 | 79 | |
BC046544.1 | - | 7 - 8 | 776 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |