Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
TC0600003373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
NM_001204906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
NM_001204906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
NM_001204907.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
NM_001204907.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
NM_023042.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026563.mm.1 |
NM_023042.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
TC0600003373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
NM_001204906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
NM_001204906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
NM_001204907.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
NM_001204907.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
NM_023042.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026564.mm.1 |
NM_023042.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
TC0600003373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
NM_001204906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
NM_001204906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
NM_001204907.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
NM_001204907.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
NM_023042.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600026562.mm.1 |
NM_023042.3 |
Refseq |