Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008171.3 | NP_032197.3 | 5 - 6 | 1767 | 74 | |
GenBank mRNA | D10651.2 | - | 2 - 3 | 1004 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |