Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009403.3 | NP_033429.1 | 1 - 2 | 437 | 94 | |
GenBank mRNA | AK030916.1 | - | 1 - 2 | 438 | 94 | |
AK157270.1 | - | 1 - 2 | 190 | 94 | ||
BC117093.1 | - | 1 - 2 | 376 | 40126412 | 94 | |
BC117097.1 | - | 1 - 2 | 376 | 40126416 | 94 | |
L09754.1 | - | 1 - 2 | 324 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |