Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172696.2 | NP_766284.2 | 21 - 22 | 3110 | 72 | |
GenBank mRNA | AK050545.1 | - | 17 - 18 | 2086 | 72 | |
AK077268.1 | - | 8 - 9 | 1392 | 72 | ||
AK147866.1 | - | 21 - 22 | 3059 | 72 | ||
BC082787.1 | - | 10 - 11 | 1503 | 72 | ||
BC141404.1 | - | 21 - 22 | 3086 | 9056017 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |