Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146145.2 | NP_666257.2 | 18 - 19 | 2872 | 72 | |
GenBank mRNA | AK141210.1 | - | 18 - 19 | 2814 | 72 | |
AK143650.1 | - | 18 - 19 | 2819 | 72 | ||
AK148521.1 | - | 18 - 19 | 2836 | 72 | ||
AK152128.1 | - | 18 - 19 | 2835 | 72 | ||
AK183753.1 | - | 3 - 4 | 346 | 72 | ||
BC024100.1 | - | 10 - 11 | 1282 | 72 | ||
BC031297.1 | - | 8 - 9 | 993 | 72 | ||
S63728.1 | - | 18 - 19 | 2822 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |