Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162925.1 | NP_001156397.1 | 6 - 7 | 826 | 63 | |
NM_030094.1 | NP_084370.1 | 6 - 7 | 775 | 63 | ||
GenBank mRNA | AK019109.1 | - | 6 - 7 | 780 | 63 | |
AK134929.1 | - | 6 - 7 | 763 | 63 | ||
AK141256.1 | - | 6 - 7 | 763 | 63 | ||
BC115559.2 | - | 6 - 7 | 811 | 40101853 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |