Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC115770.1 | - | 2 - 3 | 198 | 40085898 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |