Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100017158.mm.1 |
NM_015732.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017158.mm.1 |
NM_015732.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017158.mm.1 |
TC1100001820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017154.mm.1 |
NM_015732.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017154.mm.1 |
NM_015732.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017154.mm.1 |
TC1100001820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017159.mm.1 |
NM_015732.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017159.mm.1 |
NM_015732.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017159.mm.1 |
TC1100001820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017162.mm.1 |
NM_015732.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017162.mm.1 |
NM_015732.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017162.mm.1 |
TC1100001820.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017160.mm.1 |
NM_015732.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017160.mm.1 |
NM_015732.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017160.mm.1 |
TC1100001820.mm.1 |
MTA TC |