Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028017.1 | NP_082293.1 | 4 - 5 | 279 | 89 | |
GenBank mRNA | AK010275.1 | - | 4 - 5 | 332 | 89 | |
AK090379.1 | - | 4 - 5 | 241 | 89 | ||
AK154573.1 | - | 4 - 5 | 299 | 89 | ||
AK164149.1 | - | 4 - 5 | 274 | 89 | ||
AK167372.1 | - | 4 - 5 | 286 | 89 | ||
BC026977.1 | - | 4 - 5 | 279 | 5362684 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |