Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017394.4 | NP_059090.3 | 1 - 2 | 265 | 56 | |
GenBank mRNA | AB026688.1 | - | 1 - 2 | 179 | 56 | |
AK031867.1 | - | 1 - 2 | 233 | 56 | ||
BC027516.1 | - | 1 - 2 | 265 | 56 | ||
BC054765.1 | - | 1 - 2 | 208 | 6414759 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |