Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163126.1 | NP_001156598.1 | 8 - 9 | 856 | 99 | |
GenBank mRNA | AK030656.1 | - | 8 - 9 | 857 | 99 | |
CA316530.1 | - | 5 - 6 | 511 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |