Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168491.1 | NP_001161963.1 | 10 - 11 | 1412 | 150 | |
NM_001168492.1 | NP_001161964.1 | 10 - 11 | 1412 | 150 | ||
NM_011050.4 | NP_035180.2 | 10 - 11 | 1500 | 150 | ||
GenBank mRNA | AK134366.1 | - | 10 - 11 | 1501 | 150 | |
AK172654.1 | - | 10 - 11 | 1368 | 150 | ||
BC055739.1 | - | 10 - 11 | 1347 | 6414089 | 150 | |
CT010188.1 | - | 9 - 10 | 1210 | 150 | ||
D50465.1 | - | 9 - 10 | 1280 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |