Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010123.3 | NP_034253.3 | 7 - 8 | 1298 | 67 | |
GenBank mRNA | AK082006.1 | - | 7 - 8 | 1264 | 67 | |
AK145132.1 | - | 7 - 8 | 1306 | 67 | ||
AK145629.1 | - | 7 - 8 | 1263 | 67 | ||
AK149482.1 | - | 7 - 8 | 1251 | 67 | ||
AK161634.1 | - | 7 - 8 | 1255 | 67 | ||
AK167010.1 | - | 7 - 8 | 1265 | 67 | ||
AK172899.1 | - | 7 - 8 | 1235 | 67 | ||
U14172.1 | - | 7 - 8 | 1248 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus Inventoried Probe spans exons |