Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
NM_001253856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
NM_001253856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
NM_001253858.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
NM_001253858.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
NM_027161.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
NM_027161.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017200.mm.1 |
TC0400002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
NM_001253856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
NM_001253856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
NM_001253858.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
NM_001253858.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
NM_027161.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
NM_027161.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017202.mm.1 |
TC0400002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017205.mm.1 |
NM_001253856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017205.mm.1 |
NM_001253856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017205.mm.1 |
NM_027161.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017205.mm.1 |
NM_027161.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017205.mm.1 |
TC0400002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
NM_001253856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
NM_001253856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
NM_001253858.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
NM_001253858.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
NM_027161.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
NM_027161.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017206.mm.1 |
TC0400002076.mm.1 |
MTA TC |