Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159637.1 | NP_001153109.1 | 8 - 9 | 1019 | 65 | |
NM_138671.2 | NP_619612.2 | 8 - 9 | 918 | 65 | ||
GenBank mRNA | AK147917.1 | - | 8 - 9 | 996 | 65 | |
AK164862.1 | - | 6 - 7 | 1103 | 65 | ||
AK165510.1 | - | 8 - 9 | 1012 | 65 | ||
AK170095.1 | - | 8 - 9 | 1000 | 65 | ||
AK170608.1 | - | 8 - 9 | 1014 | 65 | ||
AK170960.1 | - | 8 - 9 | 1000 | 65 | ||
BC004012.1 | - | 8 - 9 | 906 | 3493623 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |