Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008072.2 | NP_032098.2 | 6 - 7 | 799 | 71 | |
GenBank mRNA | AF326546.1 | - | 6 - 7 | 692 | 71 | |
AF326564.1 | - | 6 - 7 | 692 | 71 | ||
BC116832.1 | - | 6 - 7 | 734 | 40126151 | 71 | |
BC116836.1 | - | 6 - 7 | 734 | 40126155 | 71 | |
S42882.1 | - | 6 - 7 | 792 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |