Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155349.1 | - | 10 - 11 | 1360 | 62 | |
AK162430.1 | - | 4 - 5 | 459 | 62 | ||
AK220168.1 | - | 8 - 9 | 1633 | 62 | ||
BC018593.1 | - | 3 - 4 | 244 | 62 | ||
BC051991.1 | - | 8 - 9 | 1275 | 62 | ||
BC088987.1 | - | 9 - 10 | 1295 | 62 | ||
BC138848.1 | - | 11 - 12 | 1231 | 8861870 | 62 | |
BC138849.1 | - | 11 - 12 | 1231 | 8861871 | 62 | |
U10115.1 | - | 10 - 11 | 1317 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |