Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256107.1 | NP_001243036.1 | 6 - 7 | 1580 | 90 | |
NM_001256108.2 | NP_001243037.2 | 7 - 8 | 1496 | 90 | ||
NM_145124.3 | NP_660106.2 | 6 - 7 | 1580 | 90 | ||
GenBank mRNA | AB072336.1 | - | 6 - 7 | 1142 | 90 | |
AK147940.1 | - | 7 - 8 | 1440 | 90 | ||
AK181990.1 | - | 95 | 90 | |||
AY974090.1 | - | 4 - 5 | 721 | 90 | ||
BC058086.1 | - | 5 - 6 | 839 | 90 | ||
BC141133.1 | - | 5 - 6 | 894 | 9055745 | 90 | |
BC145441.1 | - | 5 - 6 | 894 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |