Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053099.2 | NP_444329.2 | 4 - 5 | 4338 | 78 | |
GenBank mRNA | AB015614.1 | - | 1 - 2 | 129 | 78 | |
AK129143.1 | - | 2 - 3 | 3508 | 78 | ||
BC046423.1 | - | 3 - 4 | 3628 | 78 | ||
BC080865.1 | - | 3 - 4 | 3893 | 30531988 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |