Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014568.1 | - | 10 - 11 | 1894 | 64 | |
AK157398.1 | - | 10 - 11 | 1892 | 64 | ||
AK157956.1 | - | 11 - 12 | 1797 | 64 | ||
AK164493.1 | - | 10 - 11 | 1903 | 64 | ||
AK172043.1 | - | 10 - 11 | 1897 | 64 | ||
BC051109.1 | - | 9 - 10 | 1777 | 64 | ||
BC125432.1 | - | 10 - 11 | 1891 | 40129947 | 64 | |
BC132099.1 | - | 10 - 11 | 1891 | 40130376 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |