Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033352.3 | NP_001028524.1 | 2 - 3 | 1477 | 100 | |
GenBank mRNA | AK052203.1 | - | 2 - 3 | 1452 | 100 | |
AK154493.1 | - | 2 - 3 | 1478 | 100 | ||
AK158141.1 | - | 2 - 3 | 1453 | 100 | ||
BC060201.1 | - | 2 - 3 | 1466 | 100 | ||
BC141158.1 | - | 2 - 3 | 1451 | 9055770 | 100 | |
BC141159.1 | - | 2 - 3 | 1451 | 9055771 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |