Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039866.1 | - | 13 - 14 | 1481 | 66 | |
AK050894.1 | - | 16 - 17 | 1957 | 66 | ||
BC019659.1 | - | 7 - 8 | 787 | 66 | ||
BC050089.1 | - | 16 - 17 | 1955 | 30018732 | 66 | |
W87071.1 | - | 4 - 5 | 213 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |