Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033280.2 | NP_001028452.1 | 2 - 3 | 262 | 63 | |
GenBank mRNA | AK131907.1 | - | 2 - 3 | 263 | 63 | |
AK131908.1 | - | 2 - 3 | 263 | 63 | ||
BC147098.1 | - | 2 - 3 | 251 | 9056146 | 63 | |
BC147099.1 | - | 2 - 3 | 251 | 9056134 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |