Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013562.2 | NP_038590.2 | 10 - 11 | 1397 | 72 | |
GenBank mRNA | AK078888.1 | - | 1395 | 72 | ||
AK168682.1 | - | 10 - 11 | 1393 | 72 | ||
AK183550.1 | - | 2 - 3 | 148 | 72 | ||
AK213011.1 | - | 1 - 2 | 44 | 72 | ||
BC037013.1 | - | 10 - 11 | 1372 | 72 | ||
BC043723.1 | - | 10 - 11 | 1364 | 5369100 | 72 | |
X17400.1 | - | 10 - 11 | 1374 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |