Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042605.1 | NP_001036070.1 | 6 - 7 | 664 | 107 | |
GenBank mRNA | AK154096.1 | - | 6 - 7 | 695 | 107 | |
BC003476.1 | - | 6 - 7 | 664 | 2650401 | 107 | |
BG148172.1 | - | 4 - 5 | 332 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |