Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013797.5 | NP_038825.1 | 1 - 2 | 383 | 80 | |
GenBank mRNA | AB031813.1 | - | 1 - 2 | 224 | 80 | |
AF148218.1 | - | 1 - 2 | 207 | 80 | ||
AF223067.1 | - | 1 - 2 | 217 | 80 | ||
AK041125.1 | - | 1 - 2 | 231 | 80 | ||
AK165360.1 | - | 1 - 2 | 238 | 80 | ||
AY195868.1 | - | 1 - 2 | 216 | 80 | ||
AY243579.1 | - | 1 - 2 | 216 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |