Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007443.3 | NP_031469.1 | 3 - 4 | 419 | 68 | |
GenBank mRNA | AK004907.1 | - | 3 - 4 | 420 | 68 | |
BC021660.1 | - | 3 - 4 | 438 | 4193922 | 68 | |
D28812.1 | - | 3 - 4 | 420 | 68 | ||
X68680.1 | - | 3 - 4 | 381 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |