Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400023068.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023068.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023068.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023068.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023068.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023068.mm.1 |
NM_028640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023069.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023069.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023069.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023069.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023069.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023069.mm.1 |
NM_028640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023071.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023071.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023071.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023071.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023071.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023071.mm.1 |
NM_028640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023066.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023066.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023066.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023066.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023066.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023066.mm.1 |
NM_028640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023064.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023064.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023064.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023064.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023064.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023064.mm.1 |
NM_028640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023067.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023067.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023067.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023067.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023067.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023067.mm.1 |
NM_028640.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023070.mm.1 |
TC0400002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400023070.mm.1 |
NM_001008791.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023070.mm.1 |
NM_001008792.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023070.mm.1 |
NM_001008793.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023070.mm.1 |
NM_001276371.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400023070.mm.1 |
NM_028640.2 |
Refseq |