Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008791.2 | NP_001008791.1 | 3 - 4 | 1729 | 68 | |
NM_001008792.2 | NP_001008792.1 | 3 - 4 | 1729 | 68 | ||
NM_001008793.2 | NP_001008793.1 | 3 - 4 | 1729 | 68 | ||
NM_028640.2 | NP_082916.1 | 3 - 4 | 1729 | 68 | ||
GenBank mRNA | AK122523.1 | - | 3 - 4 | 860 | 68 | |
AK157955.1 | - | 3 - 4 | 1730 | 68 | ||
AY739114.1 | - | 3 - 4 | 1689 | 68 | ||
AY739115.1 | - | 3 - 4 | 1689 | 68 | ||
AY739116.1 | - | 3 - 4 | 1689 | 68 | ||
AY739117.1 | - | 3 - 4 | 1689 | 68 | ||
BC150757.1 | - | 3 - 4 | 1086 | 9087668 | 68 | |
HQ148552.1 | - | 3 - 4 | 971 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |