Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222993 |
NM_001007569.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222993 |
AK002994.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222993 |
BC048228.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222993 |
NM_001007569.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222993 |
AK002994.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222993 |
BC048228.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
953998 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK002515.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK010664.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK033413.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK044691.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK077153.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK077683.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK077706.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK088239.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK148337.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK151843.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK152475.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK152544.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK152603.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK153049.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK153555.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK159469.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK167823.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK168928.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK169382.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK184024.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK188100.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK197786.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK198884.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK216984.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
AK218436.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
BC005600.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
BC016455.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
BC024982.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
485339 |
NM_027352.3 |
refseq_mrna_mouse_NM |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P105626 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222993 |
NM_001007569.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222993 |
AK002994.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222993 |
BC048228.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222993 |
NM_001007569.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222993 |
AK002994.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222993 |
BC048228.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984159 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984176 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984185 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984171 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984186 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984189 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984174 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984180 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984190 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984155 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984172 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984191 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984182 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984161 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984178 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984187 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984192 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984157 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984183 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24984179 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P105626 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P222993 |
NM_001007569.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P222993 |
AK002994.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P222993 |
BC048228.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P222993 |
NM_001007569.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P222993 |
AK002994.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P222993 |
BC048228.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P105626 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06003136 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06350024 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06003128 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06003145 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06003139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984199 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984178 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984182 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24984136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05887293 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06416349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06371621 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984203 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984208 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06540150 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05925640 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05977472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984179 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984174 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984143 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05887289 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06003130 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984171 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05887295 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06367308 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06003143 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06250615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06540148 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984142 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05977470 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984155 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06412640 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984191 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984138 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984145 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06556968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984178 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984149 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984140 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984182 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984147 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06556967 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984183 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05973378 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984176 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984207 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984144 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06251355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06003142 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984172 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984141 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06350024 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984146 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06029763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984189 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24984205 |
- |
- |